missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF R alpha (Research Grade Olaratumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
Marca: R&D Systems MAB10352-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifica
| PDGFR alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 5156 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| Hu136 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Human PDGF R alpha | |
| 25 μg | |
| Primary | |
| Detects Human PDGF R alpha in direct ELISAs.This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Olaratumab. This product is for research use only. | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto