Learn More
Human Phospho-GSK-3 beta (S9) Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Marca: R&D Systems MAB8934
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifica
| GSK-3 beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2932 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1253C | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL | |
| P49840 | |
| GSK3B | |
| Phosphopeptide containing the human GSK-3 beta S9 site | |
| 100 μg | |
| Primary | |
| Detects human GSK-3 beta when phosphorylated at S9 in Western blots. | |
| Human | |
| Purified |
Titolo del prodotto
For Research Use Only