Learn More
Human RAR gamma/NR1B3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems PP-H5620-00
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
RAR gamma/NR1B3 Monoclonal specifically detects RAR gamma/NR1B3 in Human samples. It is validated for Western Blot, Immunoprecipitation, Direct ELISA.
Specifica
| RAR gamma/NR1B3 | |
| Monoclonal | |
| Unconjugated | |
| A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
| RARG | |
| Recombinant human RAR gamma/NR1B3 | |
| 100 μg | |
| Primary | |
| This antibody specifically recognizes human RAR gamma and does not cross-react with human RAR alpha or RAR beta . Not yet tested in other species. | |
| This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunoprecipitation, ELISA | |
| H5620 | |
| Western Blot, Immunoprecipitation, Direct ELISA | |
| NR1B3, NR1B3RARC, Nuclear receptor subfamily 1 group B member 3, RARC, RARG, RAR-gamma, retinoic acid receptor gamma, retinoic acid receptor, gamma | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5916 | |
| Human | |
| Purified |