missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RBP4/Retinol-Binding Protein 4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB33781-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifica
| RBP4/Retinol-Binding Protein 4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5950 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 393006 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P02753 | |
| RBP4 | |
| Mouse myeloma cell line NS0-derived recombinant human RBP4/Retinol-Binding Protein 4 Glu19-Leu201 Accession # P02753 | |
| 100 μg | |
| Primary | |
| Detects human RBP4/Retinol-Binding Protein 4 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse RBP4 is observed. | |
| Human | |
| Supernatant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto