missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Amyloid A4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF3856
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Serum Amyloid A4 Polyclonal specifically detects Serum Amyloid A4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifica
| Serum Amyloid A4 | |
| Polyclonal | |
| Unconjugated | |
| P35542 | |
| SAA4 | |
| E. coli-derived recombinant human Serum Amyloid A4 Glu19-Tyr130 Accession # P35542 | |
| 100 μg | |
| Primary | |
| Detects human Serum Amyloid A4 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Serum Amyloid A4 is observed and less than 5% cross-reactivity with recombinant human (rh) Serum Amyloid A1 and rhSerum Amyloid A2 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| C-SAA, C-SAACSAAConstitutively expressed serum amyloid A protein, SAA4, SAA5, serum amyloid A-4 protein, serum amyloid A4, constitutive | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6291 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto