missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TCL1B Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF6794
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TCL1B Polyclonal specifically detects TCL1B in Human samples. It is validated for Immunohistochemistry.
Specifica
| TCL1B | |
| Polyclonal | |
| Unconjugated | |
| O95988 | |
| TCL1B | |
| E. coli-derived recombinant human TCL1B Ala2-Asp128 (Glu93Arg) Accession # O95988 | |
| 100 μg | |
| Primary | |
| Detects human TCL1B in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human TCL1A is observed. | |
| Human | |
| IgG |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| MTCP1-like protein 1, Oncogene TCL1B, Oncogene TCL-1B, SYN-1, Syncytiotrophoblast-specific protein, T-cell leukemia/lymphoma 1B, T-cell leukemia/lymphoma protein 1B, T-cell lymphoma/leukemia 1B, TCL1, TCL1/MTCP1-like protein 1, TML1, TML1TCL1/ MTCP1-like 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9623 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto