missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TNF-alpha PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Marca: R&D Systems IC210P
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifica
| TNF-alpha | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| TNF | |
| Recombinant human TNF-alpha | |
| 100 Tests | |
| Primary | |
| Detects cytoplasmic forms of human TNF-alpha . | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 6402 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7124 | |
| Human | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto