missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total E-Cadherin DuoSet IC ELISA

Codice prodotto. p-4754528
Click to view available options
Quantità confezione:
15 Plates
2 Plates
5 Plates
Dimensione della confezione:
1 set
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30021376

Marca: R&D Systems™ DYC4225E

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Contains the basic components required for the development of sandwich ELISAs to measure in cell lysates.

The Human Total E-Cadherin DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human E-Cadherin.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantità 1 Kit
Specie Human
Coniugato Biotin
Intervallo saggio 156.00 to 10000 pg/mL
Cross-reattività <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Da utilizzare con (applicazione) ELISA
ID gene (immissione) 999
Quantità confezione 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.