missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRADD Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marca: R&D Systems MAB2658
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TRADD Monoclonal specifically detects TRADD in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifica
| TRADD | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready | |
| Q15628 | |
| TRADD | |
| E. coli-derived recombinant human TRADD Met1-Ala312 Accession # Q15628 | |
| 100 μg | |
| Primary | |
| Detects human TRADD in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BAFF R, rhTNF R1, rhHVEM, rhLT beta R, rhCD30, rhGITR, rh4-1BB, rhDR3, rhDR6, rhRANK, rhRELT, rhTAJ, rhEDAR, rhFas, recombinant mouse (rm) DC-TRAIL R1, rmDC-TRAIL R2, rhTRAIL R3, rhTRAIL R4, rhNGF R, rhCD27, rhTNF RII, rhTWEAK R, rhTRAIL R2, or rhCD40 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 313203 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC11078, TNFRSF1A-associated via death domainTNFR1-associated DEATH domain protein, tumor necrosis factor receptor type 1 associated death domain protein, tumor necrosis factor receptor type 1-associated DEATH domain protein, tumor necrosis factor receptor-1-associated protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8717 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto