missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tryptophan Hydroxylase 1/TPH-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
211.00€ - 451.00€
Specifica
| Antigene | Tryptophan Hydroxylase 1/TPH-1 |
|---|---|
| Clone | 508417 |
| Concentrazione | LYOPH |
| Diluizione | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applicazioni | Western Blot, Immunoprecipitation |
| Produktkod | Brand | Quantità | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantità | Pris | Kvantitet och tillgänglighet | |||||
18722599
![]() |
R&D Systems
MAB5276-SP |
25 μg |
211.00€
25µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
18018971
![]() |
R&D Systems
MAB5276 |
100 μg |
451.00€
100µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Beskrivning
Tryptophan Hydroxylase 1/TPH-1 Monoclonal specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifikationer
| Tryptophan Hydroxylase 1/TPH-1 | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P17752 | |
| 7166 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Tryptophan Hydroxylase 1/TPH-1 Met102-Pro403 Accession # P17752 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Tryptophan Hydroxylase 1/TPH-1 in direct ELISAs and Western blots. |
| 508417 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPH-1, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 | |
| TPH1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel