missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSC2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marca: R&D Systems AF4040
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TSC2 Polyclonal specifically detects TSC2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifica
| TSC2 | |
| Polyclonal | |
| Unconjugated | |
| P49815 | |
| TSC2 | |
| E. coli-derived recombinant human TSC2 His1506-Lys1748 Accession # P49815 | |
| 100 μg | |
| Primary | |
| Detects human TSC2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TSC1 and recombinant mouse TSC22 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| LAM, LAMFLJ43106, TSC4, Tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7249 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto