Learn More
R&D Systems™ Recombinant Human TSHR Fc Chimera Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marca: R&D Systems™ 8950-TR-050
Descrizione
The Recombinant Human TSH R Fc Chimera Protein is derived from CHO. The Recombinant Human TSH R Fc Chimera Protein has been validated for the following applications: Bioactivity.Specifica
| 7253 | |
| TSHR | |
| Chinese Hamster Ovary cell line,CHO-derived human TSH R protein, (N-terminus) Human TSH R (Met22-Gly413) Accession &Num; P16473 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
| <0.10 EU per 1μg of the protein by the LAL method. | |
| TSH R | |
| Measured by its ability to inhibit rhTSH-induced cAMP accumulation in HEK293 human embryonic kidney cells transfected with human TSH R. The ED50 for this effect is typically 2-12μg/mL. | |
| Yes |
| Theoretical M.W.: 71kDa | |
| 50μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| CHNG1, LGR3, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH R, TSH-R | |
| TSHR | |
| Unconjugated |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.