missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human USP22 (NP_056091) VersaClone cDNA
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
The Human USP22 (NP_056091) VersaClone cDNA clone contains the complete ORF for USP22, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The USP22 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifica
Specifica
| Product Type | cDNA |
| Gene | USP22 |
| Numero di accesso | NP_056091 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Tipo di inserto | 1590bp |
| ID gene (immissione) | 23326 |
| Alias gene | Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Vedi altri risultati |
For Research Use Only. Not for use in Humans.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto