Learn More
Hamilton™ HVCX Large Body Ceramic Valves
Codice prodotto. 11557884
Click to view available options
Materiale:
Aluminum
Stainless Steel
Modello:
4-5 Model
8-5 Model
Da utilizzare con:
PSD/2, 3 Syringe Drive Module
PSD/4, 8 Syringe Drive Module
Dimensione della confezione:
Pezzo
Codice prodotto. 11557884
Marca: Hamilton™ 676601
Designed to provide superior performance with salt solutions.
  • Rigid ceramic fluid path is resistant to scratching from salt crystals
  • Provides superior life and performance
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
  • Ideal for low pressure applications
Specifica
Descrizione 8-5 Model, 8 Ports, Stainless Steel Housing
Da utilizzare con PSD/2, 3 Syringe Drive Module
Materiale Stainless Steel
Modello 8-5 Model
Tipo Ceramic Large Body Valve