Learn More
Hamilton™ HVX Large Body Valves
Click to view available options
Diameter (Metric) Outer Tubing:
1.5 mm
3 mm
Dimensione della confezione:
Pezzo
Descrizione
- Physically the largest valves
- Even moderately viscous fluids flow freely
- Exceptionally easy to service and modify
Specifica
Specifica
| Materiale | Aluminum Alloy, Aluminum Housing |
| Diameter (Metric) Outer Tubing | 3 mm |
| Include | 4-4 Model, 4 Ports |
| Pressione di esercizio | 100 psig/6.9 bar |
| Tipo | Loop Flow Path |
| Temperatura massima (sistema metrico) | 38°C |