Learn More
Cytiva HyClone™ AdvanceSTEM™ Mesenchymal Stem Cell Expansion Kit
Click to view available options
Quantità:
1000 mL
500 mL
Kit - to make 11 mL complete media
Dimensione della confezione:
1 set
1000mL
500mL
Descrizione
Support expansion and maintenance of undifferentiated human mesenchymal stem cells (hMSCs) with HyClone™ AdvanceSTEM™ Mesenchymal Stem Cell Expansion Kit.
Specifica
Specifica
| Tipo di cellula | Undifferentiated Human Mesenchymal Stem Cells |
| Forma | Liquid |
| Product Type | Stem Cell Differentiation Media |
| Sterilità | Sterile-filtered |
| Quantità | 1000 mL |
| Content And Storage | Store at 2° to 8°C away from light |
| Descrizione | Mesenchymal Stem Cell Basal Medium |