Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand Origin)
Click to view available options
Quantità:
500 mL
Elaborazione:
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None
Dimensione della confezione:
500mL
Descrizione
Specifica
Specifica
| Test aggiuntivi | Progesterone,Chloride,Insulin,Inorganic Phosphorus,PTH,Glucose,Cholesterol,Copper,HDL,Magnesium,LDL,Selenium,Triglycerides,IgG,Gamma Globulin,pH,Phospholipids,Osmolality,Alkaline Phosphatase,Iron,Lactate Dehydrogenase,TIBC,SGPT,Percent Saturation,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,Total Protein,Albumin,Blood Urea Nitrogen |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C |
| Quantità | 500 mL |
| Sterilità | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm |
| Grado | Characterized |
| Elaborazione | Irradiated, 25 to 40 doses |
| Testato per | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
| Concentrazione di endotossine | ≤ 20 EU/mL |
| Calore inattivato | No |
| Vedi altri risultati |