Learn More
Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)
Codice prodotto. 16241102
Click to view available options
Quantità:
100 mL
500 mL
1000 mL
Elaborazione:
Gamma Irradiated
Heat Inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None
Dimensione della confezione:
1000mL
100mL
500mL
Codice prodotto. 16241102
Marca: Cytiva SH30087.04IR2540
Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.
  • Age at time of collection: typically 16 - 22 weeks
  • Iron-supplemented with additional growth promoting components
TRUSTED_SUSTAINABILITY
Specifica
Test aggiuntivi Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride
Certificazione/Conformità Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Additivo Iron and naturally derived components
Quantità 1000 mL
Sterilità Sterile
Sterilization Method Sequential-filtered, 100 nm, Gamma-irradiated
Elaborazione Irradiated, 25 to 40 doses
Testato per Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV
Calore inattivato No