Learn More
Cytiva HyClone™ USDA Import Tested Fetal Bovine Serum
Descrizione
- Complete traceability back to original source
- Low in antibodies and high in growth factors
- Produced using true pool technology for uniformity and consistency from bottle to bottle
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Virus panel testing according to 9 CFR 113.53
HyClone™ USDA-tested Fetal Bovine Serum (FBS) provides important proteins, hormones, growth factors, metabolites, and nutrients for your cell culture processes at a competitive price. USDA-tested FBS is ideal for typical cell culture research applications.
USDA-Tested Fetal Bovine Serum (FBS) is sourced from bovine spongiform encephalopathy (BSE)-negligible regions in throughout the world for use in low-risk applications. This serum is filtered in the US and considered to be foot and mouth disease (FMD)-free. Each lot is safety tested by the U.S. Department of Agriculture (USDA) in accordance with safety protocols required for the serum country of origin. This serum is largely sourced from Central America.
Specifica
Specifica
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
| Forma | Liquid |
| Product Type | Fetal Bovine Serum |
| Quantità | 1 L |
| Elaborazione | None |
| Concentrazione di endotossine | ≤ 25 EU/mL |
| Calore inattivato | No |
| Concentrazione emoglobina | ≤ 25 mg/dL |
| Linea di prodotti | HyClone |
| Micoplasma rilevato | Not detected |
| Vedi altri risultati |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.