Learn More
Ansell™ Hyd-Tuf™ 52-502 Series Brown Nitrile Medium Weight Gloves
Click to view available options
Size:
9
10
Dimensione della confezione:
12 paia
Descrizione
- Superior resistance to abrasion and good protection against snags, cuts and puncture
- High performance nitrile impregnation
- Excellent dry grip.
- Makes handling quicker and easier.
- Effectively repels grease, oil and dirt
- Safely machine-washable: will not shrink, stiffen or lose its shape.
- More hygienic and longer lasting – can be safely and economically re-used.
Specifica
Specifica
| Materiale parte posteriore | Nitrile |
| Certificazione/Conformità | Standard CATEGORY II EN 388 : 3111 |
| Materiale polso | Nitrile |
| Coating Material | Nitrile |
| Product Type | Medium Weight Gloves |
| Size | 9 |
| Stile | 52-502 |
| Stile polso guanto | Knit |
| Rivestimento interno | Cotton Jersey |
| Tipo di palmo | Fully coated-palm |
| Vedi altri risultati |