Learn More
Ansell HyFlex™ Medium-Duty Mechanical Protection Cut-Resistant Gloves
Click to view available options
Size:
6
7
8
9
10
11
Dimensione della confezione:
12 paia
144 paia
Descrizione
- Constructed of DSM Dyneema™, Spandex™ and nylon
- Ergnomic design promotes wearer comfort
- Special formulation of polyurethane coating for flexibility and fit provides sure grip with palm-coated finish
- Can be washed and reused without any loss of cut protection
- Strong cut resistance
- Knitwrist cuff
- Black and gray color
Compliance
ANSI™ Level 2 cut protection.
Specifica
Specifica
| ANSI Abrasion Rating | ANSI 5 |
| Certifications/Compliance (Europe) | CE, EN 420:2003 + A1:2009; Category II |
| Materiale polso | Knit |
| Coating Material | Polyurethane |
| Colore | Black, White |
| Cut Resistance Rating | ANSI A2 |
| Product Type | Cut-resistant Gloves |
| Size | 11 |
| EN388 Standard | EN 388:4342 |
| Da utilizzare con (applicazione) | Glass manufacturing, assembly, plastics injection and moulding, automotive, handling of sharp edged objects, preassembling, cutting of small dry or lightly oiled parts, maintenance |
| Vedi altri risultati |