Learn More
Ansell HyFlex™ 48-101 Polyurethane Coated Seamless Knit Nylon Gloves
Codice prodotto. p-6376000
Click to view available options
Size:
6
7
8
9
10
11
Dimensione della confezione:
12 paia
Codice prodotto. 12640922
Marca: Ansell 48101060
Solo null disponibili
Aggiungi al carrello
Edge
Aggiungi al carrello
Gloves are designed for light applications that require precise handling of small objects in dry or slightly oily environments.
  • Designed for light duty applications that require comfort and abrasion resistance
  • Polyurethane coating enhances grip in dry or light oil applications
  • Seamless construction prevents irritation
TRUSTED_SUSTAINABILITY
Specifica
ANSI Abrasion Rating ANSI 3
Chlorinated No
Area rivestita Palm
Struttura Knit
Coating Material Polyurethane
Colore Black
Cut Resistance Rating ANSI A1
Finitura esterna Rough
Hand Specific
Materiale Nylon
Product Type Seamless Knit Gloves
Size 6
Da utilizzare con (applicazione) Assembly, Inspection, and Packaging
Sesso Unisex
Stile polso guanto Knit
Grip Design Rough
Serie 48-101
Yarn Gauge 13