missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyoscine Hydrobromide Trihydrate, LoGiCal
Click to view available options
Quantità:
10 mg
Dimensione della confezione:
10mg
Specifica
Specifica
| Materiale o nome chimico | Hyoscine Hydrobromide Trihydrate |
| CAS | 6533-68-2 |
| Nomi componenti o analita | Hyoscine Hydrobromide Trihydrate |
| Shipping Condition | Room Temperature |
| Formula molecolare | C17 H21 N O4 . Br H . 3 H2 O |
| Certificazione/Conformità | ISO 9001 |
| Sinonimo | Atropine Imp. F (EP), Hyoscine Butylbromide Imp. A (EP), Hyoscyamine Sulfate Imp. D (EP), Hyoscyamine Imp. D (EP), Hyoscine Hydrobromide Trihydrate, Hyoscyamine Sulphate Imp. D (EP) as Hydrobromide Trihydrate, Hyoscine Butylbromide Imp. A (EP) as Hydrobromide Trihydrate, Atropine Imp. F (EP) as Hydrobromide Trihydrate, (alphaS)-alpha-(Hydroxymethyl)benzeneacetic acid (1alpha,2beta,4beta,5alpha,7beta)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide hydrate (1:1:3), (alphaS)-alpha-(Hydroxymethyl)benzeneacetic acid (1alpha,2beta,4beta,5alpha,7beta)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide trihydrate, alpha-(Hydroxymethyl)benzeneacetic acid [7(S)-(1alpha,2beta,4beta,5alpha,7beta)]-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide trihydrate, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane benzeneacetic acid deriv., (-)-Scopolamine hydrobromide trihydrate, Hyoscine hydrobromide trihydrate, Scopolamine hydrobromide trihydrate, Atropine Impurity F as Hydrobromide Trihydrate, Hyoscine Butylbromide Impurity A as Hydrobromide Trihydrate, Hyoscyamine Sulfate Impurity D, Hyoscyamine Sulfate Impurity D as Hydrobromide Trihydrate, Hyoscyamine Impurity D, Hyoscyamine Impurity D as Hydrobromide Trihydrate |
| InChI Formula | InChI=1S/C17H21NO4.BrH.3H2O/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;;;;/h2-6,11-16,19H,7-9H2,1H3;1H;3*1H2/t11-,12-,13-,14+,15-,16+;;;;/m1..../s1 |
| Tipo di soluzione | Neat |
| SMILES | O.O.O.Br.CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4 |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto