Learn More
Corning™ HYPERFlask™ M Cell Culture Vessel
Sfoglia Tutto Corning Prodotti
Click to view available options
:
Culture Area: 1720cm2
Descrizione
Originally, the HYPERFlask™ cell culture vessel was developed for automation. Due to its popularity for use on the bench top, Corning™ has redesigned the HYPERFlask™ M vessel with new features.
- Ergonomic Cap – smooth texture on liner cap allows more ergonomic closing and opening of vessel by operator
- Adaptor Grid – new design allows for faster filling and emptying while reducing foam generation
- Bar Code Serial Number – each individual flask is traceable by a serial number that can be read by the unaided eye or by a handheld bar code reader
- Made with USP Class VI Materials – molded and assembled in a Certified Class 100,000 clean room
- Sterile – gamma irradiated to sterility assurance level (SAL) 10−6
- Clean Room Packaging – each pack is double bagged.
- Innovative 10-Layer Design – 10 interconnected polystyrene growth surfaces
- Growth Surface – Gas permeable polystyrene
- Increase Cell Yield – 10 -fold higher cell yield increases productivity and capacity
- Time and Space Savings – Reduce processing time and incubator storage space by handling one flask compared to 10 traditional 175 cm² flasks
Specifica
Specifica
| Codice a barre | Yes |
| N. per custodia | 4/Pk. |
| N. per confezione | 24 |
| Sterilità | Sterile |