Learn More
Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Capacità (sistema metrico):
38 L
missing translation for 'unitSize'
pezzi 5
Description
- Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
- Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications:
- Removes the need for tank cleaning and helps reduce cycle times.
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
Specifications
| Capacità (sistema metrico) | 38 L |
| Capacità (sistema imperiale) | 10 gal. |
| Confezione esterna | Cardboard Cartons |
| Imballaggio interno protettivo | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Metodo di sterilizzazione | Irradiated |
| Linea di prodotti | HyPerforma |
| Film Description | CX3-9 |