Learn More
Thermo Scientific™ HyperSep™ C8 Cartridges
Codice prodotto. 10171263 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Peso del letto:
50 mg
100 mg
200 mg
500 mg
1000 mg
2000 mg
5000 mg
10 g
Capacità colonna (sistema metrico):
1 mL
3 mL
6 mL
15 mL
25 mL
75 mL
Dimensione della confezione:
pezzi 10
pezzi 100
pezzi 20
pezzi 30
pezzi 50
Codice prodotto. 10171263
Marca: Thermo Scientific™ 60108394
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 cartridges offer excellent performance.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ C8 Cartridges offer a less retentive alternative to C18 for polar and nonpolar compounds, and are well-suited for methods requiring less retention than C18. The cartridges are ideally suited for the analysis of:
  • Drugs and their metabolites in biological samples
  • Peptides in biological samples
HyperSep C8 SPE Cartridges provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Capacità colonna (sistema metrico) 6 mL
Descrizione 500 mg Bed Weight; 6 mL Column Volume
Peso del letto 500 mg
Da utilizzare con (applicazione) Screening of Drugs, Metabolites, Peptides in Biological Samples
Da utilizzare con (apparecchiatura) Thermo Scientific HyperSep-96 System
Dimensione particelle 40 to 60 μm
Product Line HyperSep
Assorbente HyperSep C8
Tipo Solid Phase Extraction Cartridge
Volume (sistema metrico) 6 mL
Quantità Pack of 30