Learn More
Thermo Scientific™ HyperSep™ Cyano Cartridges
Codice prodotto. 10121083 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Peso del letto:
50 mg
100 mg
200 mg
500 mg
1000 mg
2000 mg
5000 mg
10 g
Capacità colonna (sistema metrico):
1 mL
3 mL
6 mL
15 mL
25 mL
75 mL
Dimensione della confezione:
pezzi 10
pezzi 100
pezzi 20
pezzi 30
pezzi 50
Codice prodotto. 10121083
Marca: Thermo Scientific™ 60108747
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Get optimized isolation of polar compounds from nonpolar matrices, with these medium polarity cyano SPE cartridges.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
A medium polarity sorbent, Thermo Scientific™ HyperSep™ Cyano cartridges are less retentive than silica or diol, and are optimized for the isolation of polar compounds from nonpolar matrices. Typical applications include:
  • Retention of polar compounds from hexane and oils
  • Reversed phase extraction of moderately polar compounds
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifica
Capacità colonna (sistema metrico) 3 mL
Descrizione 200 mg Bed Weight; 3 mL Column Volume
Peso del letto 200 mg
Da utilizzare con (applicazione) Retention of Polar Compounds From Nonpolar Matrices
Da utilizzare con (apparecchiatura) Thermo Scientific HyperSep-96 System
Dimensione particelle 40 to 60 μm
Product Line HyperSep
Assorbente HyperSep Cyano
Tipo Solid Phase Extraction Cartridge
Volume (sistema metrico) 3 mL
Quantità Pack of 50