Learn More
Thermo Scientific™ Hypersil™ APS-2 Amino HPLC Columns
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Analyze sugars with this versatile amino propyl phase of Thermo Scientific™ Hypersil™ APS-2 Amino LC Columns.
Analyze sugars with this versatile amino propyl phase of Thermo Scientific™ Hypersil™ APS-2 Amino LC Columns. Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 quality guidelines. For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ LC Amino Columns as an alternative.
- In reversed phase mode, Hypersil APS-2 columns are excellent for carbohydrate analysis: extra sensitivity for sugars
- Alternative selectivity to silica when used in normal phase
Ordering Information:
Other column dimensions are available. Also available in 10μm. Please call Customer Service for more information.
Specifica
Specifica
| Formato colonna | Colonna analitica |
| Diametro (sistema metrico) | 2.1 mm |
| Da utilizzare con | IC |
| Lunghezza (sistema metrico) | 100 mm |
| Pressione max | 5800 psi (400 bar) |
| Dimensione particelle | 3 μm |
| pH | 2 to 8 |
| Dimensione del poro | 120 Å |
| Surface Area | 170 m²/g |
| Temperatura | 60 °C |
| Vedi altri risultati |