Learn More
Thermo Scientific™ Hypersil™ BDS C8 HPLC Columns
Codice prodotto. p-4523305 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
10 mm
50 mm
100 mm
125 mm
150 mm
250 mm
Diametro (sistema metrico):
2.1 mm
3 mm
4 mm
4.6 mm
Dimensione particelle:
2.4 μm
3 μm
3 µm
5 μm
5 µm
Dimensione della confezione:
Pezzo
pezzi 4
Codice prodotto. 10672854
Marca: Thermo Scientific™ 28205052130
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use as a robust, general-purpose column for QC/QA applications. They are high-quality, base-deactivated, fully endcapped, have similar C18 selectivity but slightly less retentive.
For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo ScientificTM HypersilTM BDS C8 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
For more challenging separations that require improved chromatographic performance, use Thermo ScientificTM Hypersil GOLDTM C8 columns as an alternative.
  • Excellent choice for acids, bases and neutral compounds
  • Less retentive than C18 for faster analyses
  • Robust columns with long column lifetimes
  • Very reliable for routine assays
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifica
Formato colonna Colonna analitica
Diametro (sistema metrico) 2.1 mm
Lunghezza (sistema metrico) 50 mm
Pressione max 5800 psi (400 bar)
Dimensione particelle 5 μm
pH 2 to 8
Dimensione del poro 130 Å
Surface Area 170 m²/g
Temperatura 60 °C
Tipo USP L7
Fase stazionaria C8
Product Line Hypersil BDS
Materiale di impaccamento Spherical, Fully Porous, Base-Deactivated Silica
Carico di carbonio 7%
Tipo di colonna A fase inversa
Con tappo
Fase A fase inversa