Learn More
Thermo Scientific™ Hypersil™ PREP ODS C18 HPLC Columns
Codice prodotto. p-7160180 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
8 mm
20 mm
30 mm
50 mm
100 mm
150 mm
250 mm
Diametro (sistema metrico):
2 mm
4.6 mm
10 mm
20 mm
25 mm
30 mm
50 mm
Dimensione particelle:
3 μm
5 μm
10 μm
Formato colonna:
Guard 1/16 in.
Guard 1/8 in.
Guard Column
Preparative Column, Standard
Preparative Column, Xtended Life
Dimensione della confezione:
Pezzo
Codice prodotto. 16388976
Marca: Thermo Scientific™ 40203052130
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Perfect for routine workflows, our Hypersil PREP ODS preparative LC column is a general purpose C18 used in a broad range of USP L1 applications.
Scale up your analytical method using Thermo Scientific™ Hypersil™ PREP ODS C18 reversed phase preparative HPLC Columns. Our columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. This C18 column is a general purpose column used in a broad range of applications, perfect for routine workflows.
For Routine Applications
  • C18 (USP L1) selectivity.
  • pH stable from 2 to 8.
  • Maximum temperature of 60°C.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
  • Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
  • Standard threaded hardware for column i.d. < 40mm.
  • Standard flange hardware for column i.d. ≥ 40mm.
  • Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
  • Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
  • Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
  • If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
  • Please contact your local sales representative for Customer Specific Orders.
  • Custom built columns are not eligible for returns.
TRUSTED_SUSTAINABILITY
Specifica
Formato colonna Preparative Column, Standard
Diametro (sistema metrico) 2 mm
Lunghezza (sistema metrico) 50 mm
Pressione max 5800 psi (400 bar)
Dimensione particelle 3 μm
pH 2 to 8
Dimensione del poro 130 Å
Surface Area 195 m²/g
Temperatura 60 °C
Tipo USP L1
Fase stazionaria C18
Materiale di impaccamento Spherical, Fully Porous Silica
Carico di carbonio 10%
Tipo di colonna A fase inversa
Con tappo
Fase A fase inversa