Learn More
uvex i-3 Guard Narrow Safety Glasses
Sfoglia Tutto Uvex Prodotti
Click to view available options
:
Lens Tint: Trasparente
Descrizione
General features
- Adjustable, multi-feature premium quality safety spectacle
- Robust polycarbonate lens with uvex’s proven supravision coating technology – permanent anti-fog and scratch-resistant lens
- Clip-in frame seal provides extra protection against particles, dirt and dust
- Narrow version, also available in standard size
- Metal-free
Safety features
- Certified in line with EN 166 (personal protective eyewear) and EN 170 (UV protective filter)
- Marking: W 166 FT CE – 2C-1,2 W1 FTKN CE
- UV400 – 100% protection against dangerous UVA and UVB radiation up to 400 nm
Comfort features
- Combination of soft padding around the brow with ergonomically shaped side arms and soft-grip temple ends provide a comfortable, secure fit without pressure
- Side arms with multi-incline points allow for a high level of individualization to ensure a good fit for all facial shapes and head widths
- Soft, adjustable nose pad allows spectacle to be adapted to the wearer to deliver a comfortable, non-slip and pressure-free fit
Specifica
Specifica
| Tipo di lente | Chemical-resistant |
| Lens Coating | Anti-fog, Scratch-resistant |
| Colore | Anthracite, Blue |
| Certifications/Compliance (Europe) | EN 166, EN 170 |
| Lens Tint | Clear |
| Product Type | Ergonomic Safety Glasses |
| Materiale | Polycarbonate |
| Monouso | Reusable |
| Lens Material | Policarbonato |
| Materiale del telaio | Plastic |
| Vedi altri risultati |