missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm
Click to view available options
:
60 mm plate
Descrizione
Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm is recommended for the qualitative procedures for the selective and differential isolation of B. cepacia complex in purified water systems used in the manufacture of finished products for inhalation use of non-sterile aqueous preparations for oral, oromucosal, cutaneous or nasal use.
Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm is recommended for the qualitative procedures of B. cepacia complex for water filtration testing. Confidently detect the presence of B. cepacia in nonsterile medicine with a complete workflow for USP 60 requirements which satisfies all regulations.
- Compliant formulation according to USP 60
- Quality tested to meet USP 60 specifically for Pharma customers
- Part of a complete workflow which include DCM and QCO
Specifica
Specifica
| Descrizione | Burkholderia cepacia Sel Agar (USP60) (60mm) 10/PK |
| Formato | 60 mm petri dish |
| Product Type | Agar Plate |
| Da utilizzare con (applicazione) | Pharmaceutical purified water testing used in non-sterile manufacturing. |
| Quantità | 10/Pk. |
| Status giuridico | Lab use only |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto