missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Mouse anti-Human, Clone: 2F9A8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60299-1-IG-20UL
This item is not returnable.
View return policy
Description
Intercellular adhesion molecule-1 (ICAM-1, CD54) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for lymphocyte function-associated (LFA) antigens, a beta 2 leukocyte integrin. The normal function of human ICAM-1 is to provide adhesion between endothelial cells and leukocytes after injury or stress. ICAM-1 binds to leukocyte function-associated antigen (LFA-1) or macrophage-1 antigen (Mac-1). It is found on leukocytes, fibroblasts, epithelial cells and endothelial cells and its expression is regulated by inflammatory cytokines. ICAM-1 has a tissue distribution similar to that of the major histocompatibility complex class II antigens and is likely to play a role in inflammatory responses.Specifications
| ICAM-1 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05362 | |
| ICAM1 | |
| ICAM-1 Fusion Protein Ag8309 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin) | |
| 2F9A8 | |
| Unconjugated | |
| ICAM1 | |
| BB2, CD54, ICAM 1, ICAM1, ICAM-1, P3.58 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3383 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction