Learn More
IDF-11774, MedChemExpress
Product Code. 16484438
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484438 10 mg 10mg
16430757 50 mg 50mg
16427167 5 mg 5mg
16421159 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16484438 Supplier Medchem Express Supplier No. HY111387
 Restricted Item
Spedizione stimata: 23-11-2026
MedChemExpress IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.
IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.
Specifications
Chemical Name or Material IDF-11774
CAS 1429054-28-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H32N2O2
Quantity 10 mg
Solubility Information DMSO : 60 mg/mL (162.82 mM; Need ultrasonic and warming)
SMILES CN1CCN(C(COC2=CC=C(C3(C4)CC5CC4CC(C5)C3)C=C2)=O)CC1
Molecular Weight (g/mol) 368.51
Formula Weight 368.51
Percent Purity 95.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only