missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IFN gamma ProQuantum Immunoassay Kit
GREENER_CHOICE

Codice prodotto. 16351984
Click to view available options
Quantità:
5 x 96 Tests
96 Tests
Dimensione della confezione:
Pezzo
pezzi 96
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16351984

Marca: Invitrogen™ A41150

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

ELISA

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifica

Numero di accesso P01580
Intervallo saggio 0.064 to 5,000 pg/mL
Assay Sensitivity <0.05 pg/mL
Coniugato FAM™
Metodo di rilevamento Taqman™ Real-time PCR
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Tipo di campione Plasma, Serum, Supernatant
Target Species Mouse
Da utilizzare con (applicazione) Protein Quantitation
Da utilizzare con (apparecchiatura) qPCR Instruments
ID gene (immissione) 15978
Simbolo del gene IFG; IFI
Tempo di incubazione 1 hr.
Contenuto kit Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantità 96 Tests
Status giuridico RUO
Volume del campione Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Requisiti di stoccaggio Frozen at -20°C
Bersaglio IFN gamma
Durata del test 30 mins
Tempo totale saggio 2 hrs
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato