missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGFBP5 Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA547775
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
In sandwich ELISAs, less than 3% cross-reactivity with recombinant human IGFBP-5 is observed, and less than 0.3% cross-reactivity with recombinant mouse (rm) IGF-I, rmIGF-II, rmIGFBP-1, rmIGFBP-2, rmIGFBP-3, and rmIGFBP-6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The insulin-like growth factor-binding proteins, or IGFBPs, are a family of homologous proteins that have co-evolved with the IGFs. They serve not only as shuttle molecules for the soluble IGFs, but also confer a level of regulation to the IGF signaling system. Physical association of the IGFBPs with IGF influences the bio-availability of the growth factors, as well as their concentration and distribution in the extracellular environment. In addition, the IGFBPs appear to have biological activity independent of the IGFs. Seven IGFBPs have thus far been described, each differing in their tissue distribution, half-lives and modulation of IGF interactions with their receptors. For instance, IGFBP1 is negatively regulated by insulin production. The IGFBP1 gene is expressed at a high level during fetal liver development and in response to nutritional changes and diabetes. It has been suggested that IGFBP2 functions as chaperone, escorting IGFs to their target tissues. It is expressed in several human tissues including fetal eye and fetal brain. IGFBP3 is the most abundant IGFBP and is complexed with roughly 80% of the serum IGFs. Both IGFBP3 and IGFBP4 are released by dermal fibroblasts in response to incision injury. IGFBP5 is secreted by myoblasts and may play a key role in muscle differentiation.
Specifica
| IGFBP5 | |
| Polyclonal | |
| Unconjugated | |
| IGFBP5 | |
| AI256729; AW208790; IBP 5; IBP5; IBP-5; IGF BP5; IGF-binding protein 5; IGFBP; IGFBP 5; Igfbp5; IGF-BP5; Igfbp-5; IGFBP-5P; Insulin like growth factor binding protein; insulin like growth factor binding protein 5; insulin-like growth factor binding protein 5; insulin-like growth factor binding protein-5; insulin-like growth factor-binding protein 5 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 16011 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q07079 | |
| IGFBP5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-5 Val15-Glu271. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto