Learn More
IgG(H+L) Goat anti-Rabbit, R-PE, Secondary Antibody, Proteintech
Goat anti-Rabbit Affinipure Secondary Antibody
Marca: Proteintech SA00008-2-500UL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| IgG(H+L) | |
| R-PE | |
| Goat | |
| 500 μL | |
| Rabbit | |
| Liquid |
| Immunofluorescence, Flow Cytometry | |
| 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
| The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
| Secondary | |
| Store at -20°C. Stable for one year after shipment. |