Learn More
IgG (H+L) Goat anti-Rabbit, Rhodamine, Polyclonal, Proteintech
Goat Polyclonal Antibody
Marca: Proteintech SA00007-2-200UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Immunofluorescence, Flow Cytometry.
Specifica
| IgG (H+L) | |
| Polyclonal | |
| Rhodamine | |
| 200 μL | |
| Rabbit | |
| IgG |
| Immunofluorescence, Flow Cytometry | |
| 0.6 mg/mL | |
| Goat | |
| Secondary | |
| Liquid |