Learn More
IgG (Immunoglobulin Gamma Heavy Chain) (B-Cell Marker) Mouse, Clone: rIG266, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 3500-MSM10-P0
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| IgG (Immunoglobulin Gamma Heavy Chain) (B-Cell Marker) | |
| Cytoplasm, Cell Surface and Secreted | |
| rIG266 | |
| Unconjugated | |
| Mouse | |
| 75kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Purified human Ig Gamma Chain | |
| 293T, Raji or hPBL cells. Tonsil or Spleen. | |
| 20 μg | |
| Primary | |
| IgG1 |