missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (LO-MM-9), Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals NBP1-42328
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifica
| Mouse IgM | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Mouse mu heavy chain of immunoglobulin (determined by immunodot). Avidity on IgM: 7 * 10^8 M-1. No cross reactivity with other species. | |
| Store at 4C. Do not freeze. | |
| IgG2a κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| LO-MM-9 | |
| Unconjugated | |
| PBS with 50% glycerol with 0.9% Sodium Azide | |
| IGHM | |
| The antibody reacts with the Mouse IgM antigen. | |
| 0.2 mL | |
| Primary | |
| Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto