Learn More
Proteintech IHCeasy CC2D1A Ready-To-Use IHC Kit
Codice prodotto. 18059355 Sfoglia Tutto Proteintech Prodotti
Click to view available options
Quantità:
50 Tests
Dimensione della confezione:
50 tests
Codice prodotto. 18059355
Marca: Proteintech KHC178950T
KHC1789 is a ready-to-use IHC kit for staining of CC2D1A.
KHC1789 is a ready-to-use IHC kit for staining of CC2D1A. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifica
Quantità 50 Tests