missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech IHCeasy FOXA1 Ready-To-Use IHC Kit
Sfoglia Tutto Proteintech Prodotti
Click to view available options
Quantità:
50 Tests
Dimensione della confezione:
1 set
Description
KHC0139 is a ready-to-use IHC kit for staining of FOXA1. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Spécification
Spécification
| Clonality | Polyclonal |
| Target Species | Human |
| Quantità | 50 Tests |
| Da utilizzare con (applicazione) | Immunohistochemistry (IHC) |
| Specie | Rabbit |
| Content And Storage | All the reagents are stored at 2°C to 8°C. The kit is stable for 6 months from the date of receipt. |
| Contenuto kit |
|
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu