Learn More
Proteintech IHCeasy RHAG Ready-To-Use IHC Kit
Codice prodotto. 17660137 Sfoglia Tutto Proteintech Prodotti
Click to view available options
Quantità:
50 Tests
Dimensione della confezione:
1 set
Codice prodotto. 17660137
Marca: Proteintech KHC007550T
Ready-To-Use IHC Kit
KHC0075 is a ready-to-use IHC kit for staining of RHAG. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifica
Clonality Monoclonal
Target Species Human
Quantità 50 Tests
Da utilizzare con (applicazione) Immunohistochemistry (IHC)
Specie Mouse
Content And Storage All the reagents are stored at 2°C to 8°C. The kit is stable for 6 months from the date of receipt.
Contenuto kit
  • Antigen Retrieval Buffer(100 mL, 50 x)
  • Washing Buffer(100 mL x2, 20 x)
  • Blocking Buffer(5 mL, RTU)
  • Rabbit Polyclonal Primary Antibody(5 mL, RTU)
  • Polymer-HRP-Goat anti-Mouse Secondary Antibody(5 mL, RTU)
  • Chromogen Component A(0.2 mL, RTU)
  • Chromogen Component B(4 mL, RTU)
  • Signal Enhancer(5 mL, RTU)
  • Counter Staining Reagent(5 mL, RTU)
  • Mounting Media(5 mL, RTU)
  • Datasheet (1 Copy)
  • Manual(1 Copy)