Learn More
Proteintech IHCeasy UBE3A Ready-To-Use IHC Kit
Sfoglia Tutto Proteintech Prodotti
Click to view available options
:
Quantity: 50 Tests
Descrizione
KHC1602 is a ready-to-use IHC kit for staining of UBE3A. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifica
Specifica
| Quantità | 50 Tests |
| Forma | Liquid |