Learn More
Invitrogen™ Human IL-13 ProQuantum Immunoassay Kit
Descrizione
The ProQuantum Human IL-13 Immunoassay Kit is designed to provide highly sensitive quantitative measurements of Human IL-13 protein in small sample volumes. Utilizing proximity ligation assay (PLA) technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive Cloud-based software for analytics enables sample-to-answer in just 2 hours. High sensitivity-detect low levels of protein with greater sensitivity than traditional methods Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range Small sample consumption-use 2-5 μL of sample (compared to 75 μL for triplicate wells with other methods) Fast, easy workflow-2 hours from sample to answer, with no wash steps No proprietary instrument to purchase-runs on any real-time PCR instrument ProQuantum immunoassays utilize a matched pair of target-specific antibodies, each conjugated to a DNA oligonucleotide. During antibody-analyte binding, the two DNA oligos are brought into close proximity, which allows for ligation of the two strands and subsequent creation of a template strand for amplification. This platform leverages the sensitivity and large dynamic range of Applied Biosystems TaqMan real-time...
Specifica
Specifica
| Numero di accesso | P35225 |
| Intervallo saggio | 0.0128-5000 pg/mL |
| Assay Sensitivity | <0.01 pg/mL |
| Content And Storage | Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5°C to -30°C. |
| Coniugato | FAM |
| Immunoassay Kit Format | Proximity-based Immunoassay |
| Product Type | ProQuantum |
| Tipo di campione | Plasma, Serum, Supernatant |
| Target Species | Human |
| Da utilizzare con (apparecchiatura) | qPCR Instruments |
| Vedi altri risultati |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.