Learn More
Invitrogen™ Human IL-5 ProQuantum Immunoassay Kit
Click to view available options
Quantità:
5 x 96 Tests
96 Tests
Dimensione della confezione:
96 reazioni
Pezzo
Beschreibung
IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
Spezifikation
Spezifikation
| Numero di accesso | P05113 |
| Intervallo saggio | 0.0128 to 5000 pg/mL |
| Assay Sensitivity | <0.01 pg/mL |
| Coniugato | FAM™ |
| Metodo di rilevamento | Taqman™ Real-time PCR |
| Immunoassay Kit Format | Proximity-based Immunoassay |
| Product Type | ProQuantum |
| Tipo di campione | Plasma, Serum, Supernatant |
| Target Species | Human |
| Da utilizzare con (applicazione) | Protein Quantitation |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur