missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1 beta Mouse anti-Human, Clone: 2A1B4, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifica
| Antigene | IL1 beta |
|---|---|
| Clone | 2A1B4 |
| Concentrazione | 1 mg/mL |
| Applicazioni | Western Blot, Immunohistochemistry (Paraffin) |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
16848895
|
Proteintech
66737-1-IG-20UL |
20 μL |
145.00€
20µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
16838895
|
Proteintech
66737-1-IG-150UL |
150 μL |
410.00€
150µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.Specifica
| IL1 beta | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Catabolin, IL 1, IL 1 beta, IL1 BETA, IL1B, IL1beta, IL1F2, Interleukin 1 beta, interleukin 1, beta | |
| IL1B | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2A1B4 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01584 | |
| 3553 | |
| IL1 beta Fusion Protein Ag26030 | |
| Primary | |
| -20°C | |
| IL1B |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto