missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10663-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody 10663-1-AP is raised against the full length IL18 precursor, and it recognizes both the pre-IL18 and mature IL18 theoretically.
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.Specifica
| IL18 | |
| Polyclonal | |
| Unconjugated | |
| IL18 | |
| Iboctadekin, IFN gamma inducing factor, IGIF, IL 1 gamma, IL 18, IL 1g, IL18, IL1F4, Interleukin 1 gamma, Interleukin 18, Mature IL18, pre IL18, Pre-IL18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3606 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14116 | |
| IL18 | |
| IL18 Fusion Protein Ag1063 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto