missing translation for 'onlineSavingsMsg'
Learn More

IL18R1 Mouse anti-Human, Clone: B-E43, Novus Biologicals™

Mouse Monoclonal Antibody

507.00€ - 935.00€

Specifica

Antigene IL18R1
Clone B-E43
Concentrazione 1 mg/mL
Diluizione Flow Cytometry, ELISA, Functional
Applicazioni Flow Cytometry, ELISA, Functional Assay
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
18008625
Vedere i documenti
Novus Biologicals
NBP3-14586-200UL
200 μg
507.00€
200µL
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
18329571
Vedere i documenti
Bio-Techne
NBP3-14586-500UL
500 μg
935.00€
500µL
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

IL18R1 Monoclonal antibody specifically detects IL18R1 in Human samples. It is validated for Flow Cytometry, ELISA, Functional Assay
Specifica

Specifica

IL18R1
1 mg/mL
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Mouse
CD Markers
PBS. Sterile-filtered through 0.22 um.
8809
IgG1 κ
Affinity purified
Antibody
B-E43
Flow Cytometry, ELISA, Functional
Monoclonal
Purified
RUO
Human
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Recombinant human IL18R1
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.