Learn More
Thermo Scientific™ Indiko™ and Konelab™ System Clinical Chemistry Reagents, Alkaline Phosphatase to Creatinine
Codice prodotto. p-4527895 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
15 x 5 mL
20 x 4 mL
4 x 30 mL
7 x 50 mL
8 x 20 mL
Analiti rilevabili:
ALT/GPT
AST/GOT
Alkaline Phosphatase
Creatinine (Enzymatic)
Direct Bilirubin
Dimensione della confezione:
120mL
160mL
75mL
80mL
Pezzo
Codice prodotto. 11985152
Marca: Thermo Scientific™ 981363
Standardize performance with Thermo Scientific™ Indiko™ and Konelab™ System Clinical Chemistry Reagents, which offer best-in-class methodologies.
Enhance your laboratory's performance with Thermo Scientific™ Indiko™ and Konelab™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.
Features:
  • Indiko™ and Konelab™ applications utilize carefully selected methods according to IFCC and other widely used recommendations
  • Highly developed Indiko and Konelab analyzers are integrated systems for routine clinical chemistry, ISEs, specific proteins, TDMs and DATs
  • The CE marked kits are compliant with the IVDD
  • Clinical Chemistry methods are provided with system specified multi-calibrators and -controls with traceability to international reference materials
Ordering Information: Not available in the U.S.A., except Glucose Hexokinase.
Clinical Chemistry Applications for Indiko and Konelab Analyzers:
  • Acid Phosphatase
  • Albumin
  • ALP
  • ALT
  • Amylase
  • AST
  • Bilirubin direct and total
  • Calcium
  • Cholesterol
  • HDL and LDL Cholesterol
  • Cholinesterase
  • CO2
  • Creatine Kinase
  • CK-MB
  • Creatinine
  • Gamma-GT
  • Glucose
  • alpha-HBDH
  • Iron
  • LDH
  • Lipase
  • Magnesium
  • Phosphorus
  • Total Protein
  • Triglycerides
  • U/CSF Protein
  • Urea
  • Uric Acid
TRUSTED_SUSTAINABILITY
Specifica
Analiti rilevabili AST/GOT
Tipo di campione Serum
Tipo Reagent
Quantità 7 x 50 mL
Forma Liquid